Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) chloride, ultra dry, 99.99% (metals basis)

Catalog No. p-4407472
Click to view available options
Quantity:
1 g
10 g
50 g

CAS: 7758-94-3 | Cl2Fe | 126.75 g/mol

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-94-3
Molecular Formula Cl2Fe
Molecular Weight (g/mol) 126.75
MDL Number MFCD00011004
InChI Key NMCUIPGRVMDVDB-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) dichloride
SMILES [Cl-].[Cl-].[Fe++]

Specifications

Boiling Point 1023°C
Melting Point 674°C
Color White
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,alcohol and acetone. Slightly soluble in benzene. Insoluble in ether.
Formula Weight 126.75
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Off-white powder
Vapor Pressure 13.3hPa at 700°C
Density 3.16 g/cm3 at 20°C
Chemical Name or Material Iron(II) chloride, Ultra dry
Show More Show Less
Hazard Category H302-H314-H335
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: III
EINECSNumber 231-843-4
RTECSNumber NO5400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.