Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) fumarate, 94%

Catalog No. AAB239560B
Click to view available options
Quantity:
250 g
1000 g

CAS: 141-01-5 | C4H2FeO4-2 | 169.901 g/mol

Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(II) fumarate is used as dietary Supplement, nutrient, iron source in foods and pharmaceuticals, as animal feed additive and in infant formulas.

Solubility
Soluble in water (0.14 g/100 ml at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-01-5
Molecular Formula C4H2FeO4-2
Molecular Weight (g/mol) 169.901
MDL Number MFCD00058315
InChI Key OOPLWEDSEDELIX-TYYBGVCCSA-L
Synonym ferrous fumarate, eisenfumarat
PubChem CID 66524104
IUPAC Name (E)-but-2-enedioate;iron
SMILES C(=CC(=O)[O-])C(=O)[O-].[Fe]

Specifications

Melting Point >300°C
Quantity 1000 g
Merck Index 14,4046
Solubility Information Soluble in water (0.14g/100 ml at 25°C).
Formula Weight 169.9
Percent Purity 94%
Density 2.44
Chemical Name or Material Iron(II) fumarate
EINECSNumber 205-447-7
RTECSNumber LT1950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.