Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) gluconate dihydrate, 94%

Catalog No. p-7027647
Click to view available options
Quantity:
100 g
500 g

CAS: 22830-45-1 | C12H26FeO16 | 482.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22830-45-1
Molecular Formula C12H26FeO16
Molecular Weight (g/mol) 482.17
MDL Number MFCD00150872
InChI Key OKGNXSFAYMSVNN-SYAJEJNSSA-L
Synonym iron ii gluconate hydrate
PubChem CID 131851469
SMILES O.O.[Fe++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

Specifications

Melting Point ∼140°C
Quantity 100 g
IUPAC Name iron;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;dihydrate
Formula Weight 482.19 (446.19 Anhydrous)
Percent Purity 94%
Chemical Name or Material Iron(II) gluconate dihydrate
EINECSNumber 206-076-3
RTECSNumber LZ5180000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.