Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Iron(II) gluconate hydrate, 97%

Manufacturer:  Alfa Aesar™ 8872636

Quantity 500g

 View more versions of this product

Catalog No. AA8872636


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

22830-45-1
446.19 (Anhydrous)
Iron (II) gluconate hydrate
Fe[HOCH2(CHOH)4CO2]2·xH2O (x≈2)
MFCD00150872

Specifications

22830-45-1
Fe[HOCH2(CHOH)4CO2]2·xH2O (x≈2)
MFCD00150872
500g
97%
Granular
446.19 (Anhydrous)
≈140°C
Iron (II) gluconate hydrate

Safety and Handling

SDS & Safety Documents

RTECSNumber : LZ5180000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit