Alfa Aesar™ Iron(II,III) oxide, Puratronic™, 99.997% (metals basis)

Manufacturer: Alfa Aesar™ 1296222

Quantity 100g

 View more versions of this product

Catalog No. AA1296222

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1317-61-9
    Formula Weight 231.54
    MDL Number MFCD00011010
    Merck Index 14,4040
    Chemical Name or Material Iron(II,III) oxide
    Grade Puratronic™

    Specifications

    Formula Weight 231.54
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Powder
    Refractive Index 3.0
    MDL Number MFCD00011010
    Color Black
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    CAS 1317-61-9
    Merck Index 14,4040
    Melting Point 1538°C
    Density 5.2g/mL
    TSCA TSCA
    Health Hazard 1 Warning
    Quantity 100g
    Chemical Name or Material Iron(II,III) oxide
    Assay 99.997%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit