Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) iodide, ultra dry, 99.99% (metals basis)

Catalog No. p-4407474
Click to view available options
Quantity:
1 g
10 g

Light and Air Sensitive and Hygroscopic | CAS: 7783-86-0 | FeI2

Iron(II) iodide is used as a catalyst in organic reactions. It is also used as a compound in semiconductors and other high purity applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-86-0
Molecular Formula FeI2
MDL Number MFCD00016094
Synonym iron iodide, iron ii iodide, acmc-1bdqb, iron ii iodide, anhydrous, iron ii iodide, ultra dry, iron ii iodide, anhydrous, beads,-10 mesh trace metals basis

Specifications

Color Gray
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,diethyl ether and ethanol.
Formula Weight 309.66
Odor Odorless
Sensitivity Air and light sensitive; Hygroscopic
Appearance Gray beads
Density 5.32 g/cm3
Chemical Name or Material Iron(II) iodide
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 232-031-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.