Alfa Aesar™ Iron(II) oxalate dihydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1071814

Quantity 25g

 View more versions of this product

Catalog No. AA1071814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6047-25-2
    Formula Weight 179.90 (143.87 Anhydrous)
    MDL Number MFCD00150040
    Chemical Name or Material Iron(II) oxalate dihydrate
    Grade Puratronic™

    Specifications

    Formula Weight 179.90 (143.87 Anhydrous)
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Sensitivity Hygroscopic
    MDL Number MFCD00150040
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    Physical Form Powder
    CAS 6047-25-2
    Melting Point 190°C (dec.)
    Health Hazard 1 Warning
    Density 2.28g/mL
    TSCA TSCA
    Quantity 25g
    Chemical Name or Material Iron(II) oxalate dihydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit