Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) perchlorate hydrate, Reagent Grade

Catalog No. p-7048845
Click to view available options
Quantity:
100 g
500 g

CAS: 335159-18-7 | Cl2FeO8 | 254.74 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 335159-18-7
Molecular Formula Cl2FeO8
Molecular Weight (g/mol) 254.74
MDL Number MFCD00150234
InChI Key CKFMJXZQTNRXGX-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diperchlorate
SMILES [Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Melting Point >100°C (decomposition)
Physical Form Crystalline
Quantity 100 g
UN Number UN1481
Solubility Information Soluble in alcohol and water (?98g/100 mL H2O at 0?°C).
Formula Weight 254.75 (Anhydrous)
Grade Reagent
Sensitivity Hygroscopic
Chemical Name or Material Iron(II) perchlorate hydrate
Hazard Category H272-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
EINECSNumber 237-704-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.