Learn More
Iron(II) sulfate heptahydrate, ACS, 99+%
Description
Iron(II) sulfate heptahydrate is used as a precursor to prepare other iron compounds such as a lawn conditioner and a mordant for wool dyeing. It is actively used in the manufacture of ink including iron gall ink. As a reducing agent, it participates in the reduction of chromate in cement. It is also used in industrial water treatment plants to remove phosphate. It is also used in the gold refining process to precipitate metallic gold. Woodworkers use aqueous solutions of ferrous sulfate to color maple wood a silvery hue. It finds application in the treatment of iron chlorosis, which arises due to deficiency of iron in horticulture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 64°C-6H2O |
Color | Green |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | >99% |
Solubility Information | Soluble in water. Insoluble in alcohol. |
IUPAC Name | λ²-iron(2+) heptahydrate sulfate |
Formula Weight | 278.02 (151.91 Anhydrous) |
Grade | ACS |
Sensitivity | Air Sensitive |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H336-H373 |
Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
EINECSNumber | 231-753-5 |
RTECSNumber | NO8510000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.