Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) sulfate heptahydrate, 99+%, ACS reagent

Catalog No. ac423730050
Click to view available options
Quantity:
5 g
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum

CAS: 7782-63-0 | FeH14O11S | 278.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64.0°C
Color Blue or Blue-Green or Green
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99+%
Linear Formula FeSO4·7H2O
Fieser 01,393; 04,237; 05,308
Merck Index 15,4084
Solubility Information Solubility in water: 25.6 g/100mL (20°C). Other solubilities: insoluble in alcohol
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Formula Weight 278.01
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass Bottle
Chemical Name or Material Iron(II) sulfate heptahydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
Recommended Storage due to inherent oxidation, ferric ion (Fe3+) content may increase on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.