Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric), Solstice

Catalog No. 6001937 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-37 500 g
60-019-34 1 kg
60-019-38 5 kg
60-044-731 6 x 1 kg
60-044-730 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-019-37 Supplier Solstice Supplier No. 12354500G
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.5-104.5% (manganometric)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
IUPAC Name iron(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula FeSO4 · 7H2O
Formula Weight 278.01g/mol
Percent Purity 99.5 to 104.5% (manganometric)
Grade puriss.
Packaging Plastic Bottle
Vapor Pressure 14.6mmHg (25°C)
Density 1.898g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H315-H319
EINECSNumber 231-753-5
RTECSNumber NO8510000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.