Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) sulfide, 99.98% (metals basis)

Catalog No. p-7049763
Click to view available options
Quantity:
5 g
25 g

CAS: 1317-37-9 | FeS | 87.91 g/mol

Iron(II) sulfide is used as a pigment in hair dyes and ceramics, glasses and bottles, and paint, Also used in lubricants and anodes, as a hydrogen sulfide source, and to treat exhaust gases. And it is also used as resulphurizing agent in the production of carbon, alloy and stainless types of free cutting steels. It acts as an alloying agent to improve machine ability of steel castings which are used to manufacture various steel components.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-37-9
Molecular Formula FeS
Molecular Weight (g/mol) 87.91
MDL Number MFCD00011013
InChI Key GNVXPFBEZCSHQZ-UHFFFAOYSA-N
Synonym ferrous sulfide, iron ii sulfide, iron sulfide, troilite, iron sulfuret, iron sulphide, iron monosulfide, iron protosulfide, ferrous monosulfide
PubChem CID 14828
IUPAC Name λ²-iron(2+) sulfanediide
SMILES [S--].[Fe++]

Specifications

Melting Point 1195°C
Color Brown to Black
Physical Form Powder
Quantity 5 g
Merck Index 14,4058
Solubility Information Soluble in water(0.0062g/L ),in acids with evolution of hydrogen sulfide. Insoluble in nitric acid.
Formula Weight 87.91
Percent Purity 99.98%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.74 g/mL
Assay (metals basis)
Chemical Name or Material Iron(II) sulfide
Show More Show Less
EINECSNumber 215-268-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.