Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(II) titanium oxide, 99.8+% (metals basis)

Catalog No. p-7049180
Click to view available options
Quantity:
50 g
250 g

CAS: 12022-71-8

Iron(II) titanium oxide is used as a chief source of titanium. It is also used to prepare titanium dioxide by reacting sulfuric acid to get titanium oxygen sulfate followed by hydrolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12022-71-8
MDL Number MFCD00064704
Synonym Iron(II) titanate

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 151.75
Percent Purity ≥99.8%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Iron(II) titanium oxide
EINECSNumber 234-667-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.