Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) bromide, 90+%

Catalog No. p-3586432
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

CAS: 10031-26-2 | Br3Fe | 295.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-26-2
Molecular Formula Br3Fe
Molecular Weight (g/mol) 295.56
MDL Number MFCD00016082

Specifications

Melting Point 200°C
Color Beige-Yellow to White
Physical Form Powder
Quantity 100 g
Assay Percent Range 90% min. (Iodometry)
Linear Formula FeBr3
Merck Index 15, 4045
Solubility Information Solubility in water: soluble
Formula Weight 295.56
Percent Purity ≥90%
Packaging Glass bottle
Chemical Name or Material Iron(III) bromide
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
EINECSNumber 233-089-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.