Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phthalimide, potassium derivative, 99%

Catalog No. AC170865000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

CAS: 1074-82-4 | C8H4KNO2 | 185.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-82-4
Molecular Formula C8H4KNO2
Molecular Weight (g/mol) 185.22
MDL Number MFCD00005887
InChI Key FYRHIOVKTDQVFC-UHFFFAOYSA-M
Synonym potassium phthalimide, potassium 1,3-dioxoisoindolin-2-ide, n-potassiophthalimide, phthalimide potassium salt, n-potassium phthalimide, unii-x6kka27dil, phthalimide, potassium salt, potassium phthalamide, 1h-isoindole-1,3 2h-dione, potassium salt, x6kka27dil
PubChem CID 3356745
IUPAC Name potassium;isoindol-2-ide-1,3-dione
SMILES [K+].O=C1[N-]C(=O)C2=CC=CC=C12

Specifications

Melting Point >300°C
Color Green or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Solubility Information Solubility in water: soluble in water
Formula Weight 185.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Phthalimide, potassium derivative
Show More Show Less
EINECSNumber 214-046-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.