Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) chloride hexahydrate, 97%

Catalog No. AAA162310E
Click to view available options
Quantity:
10,000 g
2500 g
500 g

CAS: 10025-77-1 | Cl3FeH12O6 | 270.29 g/mol

Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iron(III) chloride hexahydrate is used to prepare organically templated iron phosphates. It is used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin process for the production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards. It plays an important role in photoengraving and photography as an etching agent. It is useful as a mordant in dyeing and printing textiles. It is useful in the detection of phenols and phenolic derivatives; gamma-hydroxybutyric acids and also in the Trinder spot test for detecting salicylic acids.

Solubility
Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, sodium and sodium oxides and potassium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Boiling Point 280°C to 285°C
Melting Point 37°C
pH 1.8
Quantity 2500 g
Linear Formula FeCl3·6H2O
UN Number UN3260
Merck Index 14,4019
Solubility Information Soluble in water,alcohol,glycerol,methanol,ether and acetone. Insoluble in ethyl acetate.
Formula Weight 270.30 (162.21 Anhydrous)
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 1.82
Chemical Name or Material Iron(III) chloride hexahydrate
Show More Show Less
Hazard Category H302-H315-H317-H318-H373
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P314-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-729-4
RTECSNumber NO5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.