Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron(III) chloride hexahydrate, 97+%, ACS reagent
Click to view available options
Quantity:
5 g
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid

Specifications
Specifications
Boiling Point | 280.0°C to 285.0°C |
Melting Point | 37.0°C |
Color | White |
Physical Form | Crystalline Powder |
Quantity | 2.5 kg |
Assay Percent Range | 97.0 to 102% (Iodometry) |
Linear Formula | FeCl3·6H2O |
Fieser | 01,390; 02,199; 03,145; 04,236; 05,307; 06,259; 07,153; 08,228; 09,222; 15,158 |
Merck Index | 15,4046 |
Solubility Information | Solubility in water: 920g/L (20°C). Other solubilities: readily soluble in alcohol, acetone and ether |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. May cause an allergic skin reaction. Causes serious eye damage. |
Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. If skin irritation occurs: Get medical advice/attention. IF IN EYES: Rinse cautiously with wa |
RTECSNumber | NO5425000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction