Iron(III) chloride hexahydrate, ACS, 97.0-102.0%, Alfa Aesar

Manufacturer: Alfa Aesar 1249714

Quantity 25g

 View more versions of this product

Catalog No. AA1249714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10025-77-1
    Physical Form Lumps
    Assay Percent Range 97.0 to 102.0%
    Molecular Formula Cl3Fe·6H2O
    Formula Weight 270.30 (162.21 Anhydrous)
    MDL Number MFCD00149712
    Synonym Ferric chloride hexahydrate
    Chemical Name or Material Iron(III) Chloride Hexahydrate
    Grade ACS

    Specifications

    Formula Weight 270.30 (162.21 Anhydrous)
    Quantity 25g
    Color Yellow-brown
    UN Number UN3260
    Density 1.820g/cm3
    Odor Odorless
    Solubility Freely soluble in water, alcohol, ether, acetone.Soluble in water, alcohol, glycerol, methanol, ether and acetone. Insoluble in ethyl acetate.
    Appearance Yellow-Brown lump
    Health Hazard 1 H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.
    EINECS Number 231-729-4
    Physical Form Lumps
    Assay Percent Range 97.0 to 102.0%
    Molecular Formula Cl3Fe·6H2O
    MDL Number MFCD00149712
    CAS 10025-77-1
    Synonym Ferric chloride hexahydrate
    Chemical Name or Material Iron(III) Chloride Hexahydrate
    Grade ACS

    Description

    Applications:

    • Used to prepare organically templated Iron Phosphates
    • Used for sewerage treatment of industrial wastes, chloride hydrometallurgy (Silgarin) process for production of silicon), drinking water production, production of precursors for polyvinyl chloride (PVC) and copper-based metals in printed circuit boards
    • Plays important role in photoengraving and photography as an etching agent
    • Usedl as mordant in dyeing and printing textiles.
    • Also usedl in detection of phenols and phenol derivatives; gamma-hydroxybutyric acids, in Trinder spot test for detecting salicylic acids

    H314-H318-H302: Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit