Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%

Catalog No. p-7080657
Click to view available options
Quantity:
50 g
250 g
1000 g

CAS: 334024-15-6 | C6H5FeO7 | 244.94 g/mol

Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.

Solubility
Soluble in water.

Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334024-15-6
Molecular Formula C6H5FeO7
Molecular Weight (g/mol) 244.94
MDL Number MFCD00149615
InChI Key NPFOYSMITVOQOS-UHFFFAOYSA-K
Synonym ferric citrate monohydrate, iron iii citrate hydrate, unii-334s8t6lke, ferric citrate hydrate, ferric citrate usan, iron iii citrate tribasic monohydrate, citric acid iron iii salt, iron cento citrate hydrate, ksc534c4h, iron 3+ citrate hydrate
PubChem CID 22178220
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylate;iron(3+);hydrate
SMILES [Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 244.94 (Anhydrous)
Concentration or Composition (by Analyte or Components) Fe(III) 16.5 to 20%; Fe(II) max 5%
Sensitivity Light sensitive
Chemical Name or Material Iron (III) citrate hydrate
EINECSNumber 222-536-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.