Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) fluoride, anhydrous, 97% min

Catalog No. AA1148709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1148709 10 g
AA1148718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1148709 Supplier Thermo Scientific Chemicals Supplier No. 01148709

Due to product restrictions, please to purchase or view availability for this product.

CAS: 7783-50-8 | F3Fe | 112.84 g/mol

Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-50-8
Molecular Formula F3Fe
Molecular Weight (g/mol) 112.84
MDL Number MFCD00011012
InChI Key SHXXPRJOPFJRHA-UHFFFAOYSA-K
Synonym ferric fluoride, iron iii fluoride, iron fluoride, iron trifluoride, iron fluoride fef3, ferric trifluoride, fef3, hsdb 450, iron iii fluoride, anhydrous, ferric fluoride, anhydrous
PubChem CID 24552
IUPAC Name trifluoroiron
SMILES F[Fe](F)F

Specifications

Melting Point >1000° (sublimation)
Color Green
Physical Form Powder
Quantity 10 g
Assay Percent Range 97% min. (Metal basis)
Solubility Information Soluble in dilute hydrofluroic acid. Slightly soluble in water. Insoluble in alcohol,ether and benzene.
Formula Weight 112.84
Sensitivity Hygroscopic
Appearance Pale green powder
Density 3.87 g/cm3
Chemical Name or Material Iron(III) fluoride
Show More Show Less
Hazard Category H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.
EINECSNumber 232-002-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.