Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) fluoride trihydrate

Catalog No. p-7049157
Click to view available options
Quantity:
50 g
250 g

CAS: 15469-38-2

The primary commercial use of iron(III) fluoride in the production of ceramics. Some cross coupling reaction are catalyzed by ferric fluoride-based compounds. Iron(III) fluoride has also been shown to catalyze chemoselective addition of cyanide to aldehydes to give the cyanohydrins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15469-38-2
MDL Number MFCD00149718
Synonym iron 3+ trifluoride trihydrate, acmc-20akpl, ferric iron trihydrate trifluoride, iron 3+ ion trihydrate trifluoride, iron 3+ fluoride-water 1/3/3, fluoride, fluoride, iron salt, hydrate, hydrate, hydrate, fluoride

Specifications

Color White to Brown
Physical Form Crystalline
Quantity 50 g
UN Number UN3260
Merck Index 14,4022
Solubility Information Slightly soluble in water. Sparingly soluble in alcohol,ether,benzene.
Formula Weight 166.89 (112.84 Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Iron(III) fluoride trihydrate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 232-002-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.