Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) hydroxide, 20% in H{2}O, nanoparticle dispersion, low pH, <5.0nm APS Number Weighted

Catalog No. AA4724136
Click to view available options
Quantity:
100 g
500 g

CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide is used in cosmetics, tattoo inks and aquarium water treatment. It is used as a pigment and as an adsorbent for removing lead from aquatic media. It acts as a catalyst for proteomics research. It is involved in the removal of toxic selenate and selenite ions from water systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Color Red to Brown
pH 3.0
Physical Form Liquid
Quantity 500 g
Linear Formula FeO(OH)
Merck Index 14,4025
Solubility Information Insoluble in water.
Formula Weight 88.85
Odor Odorless
Density 1.20 g/mL
Chemical Name or Material Iron(III) hydroxide, 20% in H2O, nanoparticle dispersion
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H335
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P304+P340-P362-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Take off contaminated clothing and wash before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 243-746-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.