Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) hydroxide, gamma-phase

Catalog No. AA1753118
Click to view available options
Quantity:
2 g
10 g
50 g

CAS: 20344-49-4 | FeHO2 | 88.851 g/mol

Iron(III) hydroxide, γ-phase is used in purifying water and as an absorbent in chemical processes. It is involved in the preparation of iron oxide-hydroxides nanoparticles, which is used as a very good adsorbent for lead removal from aquatic media. It is also used as a pigment, as a catalyst and in aquarium water treatment as a phosphate binder. Further, it is employed in the paints-lacquers-varnishes industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20344-49-4
Molecular Formula FeHO2
Molecular Weight (g/mol) 88.851
MDL Number MFCD00064782
InChI Key AEIXRCIKZIZYPM-UHFFFAOYSA-M
Synonym hydroxy oxo iron, goethite, lepidocrocite, iron hydroxide oxide, goethite fe oh o, iron hydroxide oxide fe oh o, iron iii oxide-hydroxide, ferric acid, iron pigment yellow, ferric hydroxide oxide
PubChem CID 91502
IUPAC Name hydroxy(oxo)iron
SMILES O[Fe]=O

Specifications

Melting Point 135°C (decomposition)
Color Brown
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 88.85
Odor Odorless
Chemical Name or Material Iron(III) hydroxide, Gamma-phase
EINECSNumber 243-746-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.