Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) isopropoxide, 2.5% w/v in isopropanol

Catalog No. p-7058684
Click to view available options
Quantity:
5 mL
25 mL

CAS: 14995-22-3 | C9H21FeO3 | 233.109 g/mol

Iron(III) isopropoxide is used in nano-materials & thin films, engineering & environmental, catalysis & synthesis and energy conversion & storage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14995-22-3
Molecular Formula C9H21FeO3
Molecular Weight (g/mol) 233.109
MDL Number MFCD00070436
InChI Key QUHDSMAREWXWFM-UHFFFAOYSA-N
Synonym iron 3+ ;propan-2-olate, iron iii i-propoxide, triisopropoxyiron iii, 2-propanol, iron 3+ salt 9ci, iron 3+ ion tris propan-2-olate, iron 3+ tris propan-2-olate, iron iii i-propoxide 99.9+%-fe
PubChem CID 11961763
IUPAC Name iron(3+);propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Fe+3]

Specifications

Color Red to Brown
Density 0.79 g/mL
Boiling Point 82°C
Flash Point 12°C (54°F)
Odor Alcohol-like
Quantity 5 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 233.11
Concentration or Composition (by Analyte or Components) 2.5% w/v in isopropanol
Physical Form Liquid
Chemical Name or Material Iron (III) isopropoxide
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.