Iron (III) Nitrate Hydrate, 99.99%, metal basis, Alfa Aesar

Manufacturer: Alfa Aesar 1071506

Quantity 5g

 View more versions of this product

Catalog No. AA1071506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-61-8
    Physical Form Crystalline Aggregates
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula Fe(NO3)3·xH2O (x ≈ 9)
    Formula Weight 241.86
    MDL Number MFCD00149708
    Chemical Name or Material Iron(III) nitrate hydrate

    Specifications

    Formula Weight 241.86
    Quantity 5g
    Color Pale purple
    Density 1.684g/cm3
    EINECS Number 233-899-5
    Physical Form Crystalline Aggregates
    Odor Acidic
    Assay Percent Range 99.999% (Metal basis)
    Molecular Formula Fe(NO3)3·xH2O (x ≈ 9)
    Appearance Pale purple crystalline aggregates
    MDL Number MFCD00149708
    Health Hazard 1 H272-H315-H319: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation.
    CAS 7782-61-8
    Chemical Name or Material Iron(III) nitrate hydrate

    H272-H315-H319: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit