Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) nitrate nonahydrate, 98+% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 12226A1

Quantity 1kg

 View more versions of this product

Catalog No. AA12226A1


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7782-61-8
Fe(NO3)3·9H2O
MFCD00149708
14,4027
FeN3O9·9H2O
403.99 (241.86 Anhydrous)
UN1466
Iron(III) nitrate nonahydrate

Specifications

7782-61-8
FeN3O9·9H2O
403.99 (241.86 Anhydrous)
UN1466
1.684
Moisture Sensitive
Pale Purple
1kg
≥98%
Crystalline, Contains Excess Hno3
Fe(NO3)3·9H2O
MFCD00149708
14,4027
Acidic
125°C (dec.)
47°C
Iron(III) nitrate nonahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H272-H315-H319;
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P221-P210-P220-P280-P305+P351+P338-P501a;
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep/Store away from clothing/combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: FERRIC NITRATE

RTECSNumber : NO7175000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit