Learn More
Iron(III) oxalate hexahydrate
Description
Iron(III) oxalate hexahydrate is used as a catalyst for the reductive and oxidative degradation of iopamidol under UV and visible light irradiation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 25 g |
UN Number | UN3288 |
Solubility Information | Soluble in water. |
Formula Weight | 483.84 (375.55 Anhydrous) |
Physical Form | Powder |
Chemical Name or Material | Iron(III) oxalate hexahydrate |
Safety and Handling
Hazard Category | H302+H312 |
Hazard Statement | GHS H Statement H302-H312 Harmful if swallowed. Harmful in contact with skin. |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S. |
EINECSNumber | 220-951-7 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.