Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) oxide, 98% (metals basis), Thermo Scientific Chemicals

Catalog No. AA12375A7
Click to view available options
Quantity:
1 kg
5 kg
25 kg

It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Melting Point 1565°C
Color Red to Brown
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98%
Solubility Information Insoluble in water,Soluble in diluted acids,sugar solution and slightly soluble in aq. tartaric acid,citric acid and acetic acid.
Formula Weight 159.69
Appearance Red-brown powder
Density 5.24 g/cm3
Chemical Name or Material Iron(III) oxide
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber 215-168-2
RTECSNumber NO7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.