Learn More
Iron(III) sulfate hydrate, Reagent Grade
Description
Iron(III) sulfate is also used in pigments, pickling baths for aluminum and steel. It acts as a mordant in dyeing, as a coagulant for industrial wastes. In the medical field, it is used as an astringent and styptic. It is involved in the preparation of aromatic esters from the corresponding acids and alcohols. Further, it is employed in the oxidation of pyrite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Color | Gray-White |
Physical Form | Granules, Powder |
Quantity | 1 kg |
Solubility Information | Soluble in waterSoluble in water. Sparingly soluble in alcohol. |
IUPAC Name | diiron(3+) hydrate trisulfate |
Formula Weight | 399.87 (Anhydrous) |
Grade | Reagent |
Odor | Odorless |
Sensitivity | Hygroscopic |
Appearance | Grayish-white powder or granules |
Show More |
Safety and Handling
Hazard Category | H302-H315-H318-H350 |
Precautionary Statement | P201-P202-P264b-P270-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 8; Packaging Group: III |
EINECSNumber | 233-072-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.