Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iron Indicator Solution 2% w/v Salicylic Acid in Methanol, Reagents
Click to view available options
Quantity:
4 L
Specifications
Specifications
Chemical Name or Material | Iron Indicator Solution 2% w/v Salicylic Acid in Methanol |
Boiling Point | 64.5°C |
CAS | 67-56-1, 69-72-7 |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 0.8 |
Color | Colorless |
Quantity | 4 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction