Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Metal, Electrolytic, Granular, Primary Standard, Approx .99.90-100%, Spectrum™ Chemical
SDP

Catalog No. 18603510 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-510 100 g Amber Glass Bottle
18-603-509 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-603-510 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. I1197100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

I1197, 7439-89-6, Fe

Spectrum™ Chemical Iron Metal, Electrolytic, Granular, Primary Standard

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
MDL Number MFCD00010999
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
IUPAC Name iron
SMILES [Fe]

Specifications

CAS Max % 100%
Quantity 100 g
Packaging Amber Glass Bottle
Form Solid
Percent Purity ∼99.9 to 100%
Grade ACS
Chemical Name or Material Iron Metal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.