Learn More
Sigma Aldrich Iron oxide incorporated conjugated polymer nanoparticles
Catalog No. 905038250UL Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 μL
Catalog No. 90-503-8250UL
Supplier: Sigma Aldrich 905038250UL
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
The 'core-shell' structure consisting of polymer and iron oxide in the core and the biocompatible surfactant in the surrounding shell allows easy covalently bonding of these nanoparticles with biomolecules such streptavidin, antibodies, proteins, nucleic acids, etc and make them suitable for the following applications:Fluorescence Microscopy (high content screening, live cell tracking and 3-D cell imaging)Immunohistochemistry and ImmunocytochemistryFlow CytometryRapid Diagnostic Tests (RDTs) Tissue Imaging2-Photon ImagingNIR Imaging
Specifications
Quantity 250 μL
Synonym Conjugated polymer nanoparticles; Fluorescent magnetic nanoparticles; Fluorescent nanoparticles; Magneto-conjugated polymer nanoparticles
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.