Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron powder, 98% (metals basis), Thermo Scientific Chemicals

Catalog No. aa3981330
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3981330 250 g
AA3981336 500 g
AA39813A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3981330 Supplier Thermo Scientific Chemicals Supplier No. 03981330

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-89-6
Molecular Formula Fe
Molecular Weight (g/mol) 55.85
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
Synonym ferrum, iron, elemental, ferrous, remko, armco, ferrovac, hoeganaes eh, 3zhp, ancor b, atomiron 5m
PubChem CID 23925
ChEBI CHEBI:82664
IUPAC Name iron
SMILES [Fe]

Specifications

Color Gray
Appearance Gray powder
Density 7.87g/cm3 at 20°C
Quantity 250 g
Mesh Size -325 Mesh
Form Powder
Assay Percent Range 98% (Metals basis)
MDL Number MFCD00010999
Chemical Name or Material Iron, Reduced
Hazard Category H500-X
Precautionary Statement P235+P410-P280
DOTInformation DOT Class: 4.2; Packaging Group: II
EINECSNumber 231-096-4
RTECSNumber NO4565500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.