Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron silicide, 99.9% (metals basis)

Catalog No. p-4407465
Click to view available options
Quantity:
25 g
100 g

CAS: 12022-95-6 | FeSi | 83.93 g/mol

Iron silicide is used in deoxidation and alloying of steel melts, alloying of cast iron charges and melts (Iron foundries), silicothermic reduction of precious metals (chromium, molybdenum and vanadium) and others (Nb, W), Production of some inoculants and nodulisers by alloying (melting). High purity iron silicide is used to produce high permeability steel for electric transformers, production of FeSiMn, FeSiN and SiMn- alloys. It is also used as metal filler.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12022-95-6
Molecular Formula FeSi
Molecular Weight (g/mol) 83.93
MDL Number MFCD00016086
InChI Key YSWDZWXHBYTMBR-UHFFFAOYSA-N
Synonym silyliron, silylene iron, ferric silicon, silanidylidyneironylium
PubChem CID 92027036
SMILES [Si-]#[Fe+]

Specifications

Melting Point 1410°C
Color Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (Metal basis)
Solubility Information Insoluble in water.
IUPAC Name silanidylidyneironylium
Formula Weight 83.93
Odor Odorless
Appearance Gray powder
Density 6.1g/cm3
Chemical Name or Material Iron silicide
Show More Show Less
Hazard Category H315-H319: Causes skin irritation. Causes serious eye irritation.
EINECSNumber 234-671-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.