Learn More
Iron silicide, 99.9% (metals basis)
Description
Iron silicide is used in deoxidation and alloying of steel melts, alloying of cast iron charges and melts (Iron foundries), silicothermic reduction of precious metals (chromium, molybdenum and vanadium) and others (Nb, W), Production of some inoculants and nodulisers by alloying (melting). High purity iron silicide is used to produce high permeability steel for electric transformers, production of FeSiMn, FeSiN and SiMn- alloys. It is also used as metal filler.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1410°C |
| Color | Gray |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99.9% (Metal basis) |
| Solubility Information | Insoluble in water. |
| IUPAC Name | silanidylidyneiron(1+) |
| Formula Weight | 83.93 |
| Odor | Odorless |
| Appearance | Gray powder |
| Show More |
Safety and Handling
| Hazard Category | H315-H319: Causes skin irritation. Causes serious eye irritation. |
| EINECSNumber | 234-671-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.