Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Standard, 1 ppm Fe, Reagents

Catalog No. 01334949
Click to view available options
Quantity:
500 mL
10 L

Iron Standard, 1 ppm Fe, Reagent, use for Iron analysis. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Iron Standard, 1 ppm Fe
Packaging Cubitainer
CAS 7732-18-5, 7697-37-2
Specific Gravity 1.02
Physical Form Liquid
Color Colorless to Slight Yellow
pH <1
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Industry Type Iron Analysis
Quantity 10 L
Grade Reagent
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.