Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Standard Solution, 50ppm (50μg/mL), LabChem™

Catalog No. LC157301 Shop All LabChem Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC157301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC157301 Supplier LabChem, Inc. Supplier No. LC157301
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7783-85-9 , 7664-93-9 , 7732-18-5
Molecular Formula FeH20N2O14S2
Molecular Weight (g/mol) 392.13
MDL Number MFCD00150530
InChI Key MQLVWQSVRZVNIP-UHFFFAOYSA-L
IUPAC Name λ²-iron(2+) diammonium hexahydrate disulfate
SMILES [NH4+].[NH4+].O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Packaging Poly Bottle
CAS Max % 99.91, 0.05, 0.04
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Concentration 50 ppm (50 μg/mL)
Linear Formula Fe(NH4)2(SO4)2·6H2O
Color Colorless
Solubility Soluble in water
Quantity 500 mL
Formula Weight 392.14
Chemical Name or Material Iron Standard Solution, 50ppm (50μg/mL)
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.