Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron standard solution, for AAS, 1 mg/ml Fe in 2 to 5% HNO3

Catalog No. p-220912
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7439-89-6 | Fe | 55.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iron standard solution
Name Note 1 mg/ml Fe in 2% HNO3
Packaging Plastic bottle
CAS 7439-89-6, 7697-37-2, 7732-18-5
Density 7.86
Physical Form Liquid
Molecular Formula Fe
Concentration 997 to 1003 μg/mL of Fe (3+) in 2% HNO3 (20°C)
MDL Number MFCD00010999
Color Colorless
InChI Key XEEYBQQBJWHFJM-UHFFFAOYSA-N
SMILES [Fe]
IUPAC Name iron
Molecular Weight (g/mol) 55.85
Quantity 100 mL
PubChem CID 23925
ChEBI CHEBI:82664
Formula Weight 55.85
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.