Iron sulfide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1284203

Quantity 1g

 View more versions of this product

Catalog No. AA1284203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12068-85-8
    Physical Form Powder
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula FeS2
    Formula Weight 119.98
    MDL Number MFCD00064690
    Chemical Name or Material Iron sulfide

    Specifications

    Formula Weight 119.98
    Quantity 1g
    Color Gray
    EINECS Number 235-106-8
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.99% (Metal basis)
    Molecular Formula FeS2
    Appearance Gray powder
    MDL Number MFCD00064690
    CAS 12068-85-8
    Chemical Name or Material Iron sulfide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit