Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Iron(III) Phosphate Dihydrate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 g
1 kg
Description
Iron(III) phosphate dihydrate (FePO4 x 2H2O) can be used as a catalyst in the synthesis of: 3,4-dihydropyrimidin-2(1H)-ones and thiones by reacting with aldehydes, +a-ketoesters and urea/thiourea via one pot-three component Biginelli reaction.Methyl methacryalate (MMA) by oxidative dehydrogenation of methyl iso-butarate (MIB). It can be also used in the synthesis of carbon coated lithium iron phosphate (LiFePO4) as the cathode material for lithium ion batteries. It is also used as a key ingredient in preparing phosphate glass fibers.
Specifications
Specifications
| CAS | 13463-10-0 |
| Molecular Formula | FeO4P -+ 2H2O |
| Linear Formula | FePO4 -+ 2H2O |
| MDL Number | MFCD00221523 |
| Quantity | 1 kg |
| Synonym | Ferric phosphate dihydrate |
| Molecular Weight (g/mol) | 186.85 g/mol |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction