Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) Phosphate Dihydrate

Catalog No. 436011250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
250 g
1 kg

Catalog No. 43-601-1250G

Supplier: Sigma Aldrich 436011250G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(III) phosphate dihydrate (FePO4 x 2H2O) can be used as a catalyst in the synthesis of: 3,4-dihydropyrimidin-2(1H)-ones and thiones by reacting with aldehydes, +a-ketoesters and urea/thiourea via one pot-three component Biginelli reaction.Methyl methacryalate (MMA) by oxidative dehydrogenation of methyl iso-butarate (MIB). It can be also used in the synthesis of carbon coated lithium iron phosphate (LiFePO4) as the cathode material for lithium ion batteries. It is also used as a key ingredient in preparing phosphate glass fibers.

Specifications

CAS 13463-10-0
Molecular Formula FeO4P -+ 2H2O
Linear Formula FePO4 -+ 2H2O
MDL Number MFCD00221523
Quantity 250 g
Synonym Ferric phosphate dihydrate
Molecular Weight (g/mol) 186.85 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.