Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iroquois-class homeodomain protein IRX-5 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | Iroquois-class homeodomain protein IRX-5 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Iroquois-class homeodomain protein IRX-5 Polyclonal specifically detects Iroquois-class homeodomain protein IRX-5 in Mouse samples. It is validated for Western Blot.Specifications
Iroquois-class homeodomain protein IRX-5 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
Homeodomain protein IRX-2A, Homeodomain protein IRXB2, iroquois homeobox 5, Iroquois homeobox protein 5, iroquois-class homeodomain protein IRX-5, IRX-2a, IRX2A, IRXB2 | |
The immunogen is a synthetic peptide directed towards the middle region of mouse Iroquois-class homeodomain protein IRX-5 (NP_061296). Peptide sequence ENIDLEKNDEDEPQKPEDKGDLEGPESGGAEQKATAGCERLQGPLSPAGK | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
10265 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title