Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRP2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | IRP2 |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
IRP2 Polyclonal specifically detects IRP2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
IRP2 | |
Polyclonal | |
Purified | |
RUO | |
Human | |
ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human IRP2 (NP_004127.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
HIF Target Genes, Hypoxia, Lipid and Metabolism | |
PBS with 50% glycerol, pH7.3. | |
3658 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title