Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS-2 Polyclonal Antibody, Abbomax

Catalog No. 502204092
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-4092 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-4092 Supplier Abbomax Inc Supplier No. 602630
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: Human kidney Cellular location: Cytoplasm.

IRS-2 Polyclonal antibody specifically detects IRS-2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen IRS-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene IRS2
Gene Accession No. P81122, Q9Y4H2
Gene Alias 4PS; insulin receptor substrate 2; Irs2; IRS-2; tyrosine kinase substrate
Host Species Rabbit
Immunogen A synthetic peptide corresponding to the epitope ASSPA without phosphorylation site Ser 731 of human IRS-2
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29376, 384783, 8660
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.