Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP222461
Description
IRS1 Polyclonal specifically detects IRS1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IRS1 | |
Polyclonal | |
Western Blot 2 μg/mL, Flow Cytometry 3 - 5 μg/ 10^6 cells, Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL | |
HIRS-1, insulin receptor substrate 1, IRS-1 | |
Rabbit | |
Affinity Purified | |
RUO | |
3667 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
PBS and 1 mg/ml BSA. with 0.05% Sodium Azide | |
IRS1 | |
Synthetic peptide corresponding to residues D(631) Y M P M S P K S V S A P Q Q I(646) of human IRS-1. | |
100 μg | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction