Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRS1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$199.93 - $537.28

Specifications

Antigen IRS1
Concentration 1.0 mg/mL
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB017854
SDP
View Documents
Novus Biologicals
NB100820010.025ML
0.025 mL
Each for $199.93
Only null left
Add to Cart
 
NB10082001
SDP
View Documents
Novus Biologicals
NB10082001
0.1 mL
Each for $537.28
Only null left
Add to Cart
 
Description

Description

IRS1 Polyclonal specifically detects IRS1 in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

IRS1
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:100-1:200, Immunohistochemistry-Paraffin 1:50-1:100
Polyclonal
Rabbit
Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Signal Transduction
P35568
3667
IgG
Affinity Purified
180 kDa
1.0 mg/mL
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Rat, Human, Mouse
HIRS-1, insulin receptor substrate 1, IRS-1
IRS1
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.