Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Antibody (OTI3G10), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271049F
Description
IRS1 Monoclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
IRS1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
IRS1 | |
Recombinant protein fragment corresponding to amino acids 1-280 of human IRS1 (NP_005535) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI3G10 | |
Immunohistochemistry | |
HIRS-1, insulin receptor substrate 1, IRS-1 | |
Mouse | |
Affinity Purified | |
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction | |
3667.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction