Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Antibody (OTI3G10), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271049PCP
Description
IRS1 Monoclonal antibody specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
IRS1 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
OTI3G10 | |
Western Blot, Immunohistochemistry | |
HIRS-1, insulin receptor substrate 1, IRS-1 | |
Human recombinant protein fragment corresponding to amino acids 1-280 of human IRS1(NP_005535) produced in E.coli. | |
0.1 mL | |
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Neuroscience, Phospho Specific, Signal Transduction | |
3667 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction