Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1 Mouse anti-Human, Clone: HIR-B1, Novus Biologicals™

Mouse Monoclonal Antibody
$220.00 - $524.00
Specifications
Antigen | IRS1 (p Ser616) |
---|---|
Clone | HIR-B1 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot, ELISA |
Applications | ELISA, Western Blot |
Description
IRS1 Monoclonal specifically detects IRS1 in Human samples. It is validated for Western Blot, ELISA.Specifications
IRS1 (p Ser616) | |
1.0 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
RUO | |
Human | |
HIRS-1, insulin receptor substrate 1, IRS-1 | |
IRS1 | |
IgG1 | |
Affinity Purified |
HIR-B1 | |
Western Blot, ELISA | |
Monoclonal | |
Mouse | |
Autophagy, Cancer, Diabetes Research, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction | |
P35568 | |
3667 | |
Synthetic peptide phosphorylated at Ser 616 site | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title