Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRS1, p Ser307 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178078
Description
IRS1 Polyclonal specifically detects IRS1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IRS1 (p Ser307) | |
Polyclonal | |
Unconjugated | |
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
HIRS-1, insulin receptor substrate 1, IRS-1 | |
Rabbit | |
131.59 kDa | |
0.1 mg | |
Autophagy, Cancer, Diabetes Research, Lipid and Metabolism, mTOR Pathway, Nutrient Sensing in the Brain, Phospho Specific, Signal Transduction | |
3667 | |
Human, Mouse, Rat, Monkey | |
IgG |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1:250-1:1500, ELISA 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
P35568, P35568, P35568, P35568 | |
IRS1 | |
This affinity purified IRS1 [p Ser307] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near amino acids 290-320 of human IRS1 protein. (Uniprot: P35568) | |
Affinity Purified | |
RUO | |
Primary | |
This affinity-purified antibody is directed against the phosphorylated form of human IRS1 protein at the pS307 residue. The resultant affinity purified antibody was then cross-adsorbed against the non-phosphorylated form of the immunizing peptide. Reactivity occurs against human IRS1 pS307 protein and the antibody is specific for the phosphorylated form of the protein. Reactivity with non-phosphorylated human IRS1 is minimal by ELISA. A BLAST analysis was used to suggest cross reactivity with IRS1 from human, mouse, rat, dog and vervet monkey based on 100% homology with the immunizing sequence. Partial reactivity is also expected against IRS1 from pig (94%), bovine (94%) and chicken (88%) sources. Reactivity of this antibody with IRS1 from other species is unknown. | |
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction